Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291741_m1
          See other LOC328066 GE Assays ›
          Gene Symbol:
          LOC328066
          Gene Name:
          uncharacterized LOC328066
          Gene Aliases:
          C920021A13
          Chromosome Location:
          Chr.12: 25316852 - 25471867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291741_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC328066
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK083360.1 - 513 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB654288.1 - 3 - 4 513 Chr. 12 - 25316190 - 25385320 [+] on Build GRCm38 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          17746
          Gm17746 -244.6kb Sense Chr. 12 - 25129174 - 25132274 on NCBI Build GRCm38
          Downstream predicted gene,
          29687
          Gm29687 706.6kb Anti-sense Chr. 12 - 26091962 - 26210505 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2314229 AK083360.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414938 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113169 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07563662 - -
          Whole Mouse Genome G4122A A_52_P247658 AK083360.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10034501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10098720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10086299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10226570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10034509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10097155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10160888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10407248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10125901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28295276 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1441600_PM_at - -
          Mouse Genome 430 2.0 Array 1441600_PM_at - -
          Mouse Genome 430 2.0 Array 1441600_at - -
          Mouse Expression Set 430 1441600_at - -
          Mouse Expression Set 430 1441600_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441600_at - -
          Affymetrix HT Mouse Genome 430B Array 1441600_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 779063 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103140022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix