Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291807_m1
          See other PTGS2OS GE Assays ›
          Gene Symbol:
          Ptgs2os
          Gene Name:
          prostaglandin-endoperoxide synthase 2, opposite strand
          Gene Aliases:
          7530420F21Rik
          Chromosome Location:
          Chr.1: 150074873 - 150099874 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291807_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptgs2os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015466.3 3 - 4 755 68
          GenBank mRNA AK083274.1 - 3 - 4 699 68
          AK144829.1 - 3 - 4 648 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phospholipase A2,
          group IVA (cytosolic,
          calcium-dependent)
          Pla2g4a Pla2g4,
          cPLA2,
          cPLA2-alpha,
          cPLA2alpha
          -123.3kb Sense Chr. 1 - 149829618 - 149961284 on NCBI Build GRCm38
          Downstream prostaglandin-endoperoxide synthase 2 Ptgs2 COX2,
          Cox-2,
          PGHS-2,
          PHS-2,
          Pghs2,
          TIS10
          12.8kb Anti-sense Chr. 1 - 150100124 - 150108017 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          negative regulation of toll-like receptor signaling pathway
          protein binding

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101472 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158051 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00213748 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158051 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04303175 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101905 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00037670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00358634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20707184 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101980324 - -
          MouseWG-6 v2 Expression BeadChip 105700047 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0