Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292065_m1
          See other 2610100L16RIK GE Assays ›
          Gene Symbol:
          2610100L16Rik
          Gene Name:
          RIKEN cDNA 2610100L16 gene
          Gene Aliases:
          100040120, Gm2612
          Chromosome Location:
          Chr.3: 17789921 - 17800071 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292065_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610100L16Rik
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033490.1
          2 - 3 683 149
          GenBank mRNA AK011787.1 - LIT3367
          2610100L16
          2 - 3 658 149
          AK163433.1 -
          2 - 3 556 149

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          30340
          Gm30340 -8.6kb Anti-sense Chr. 3 - 17287413 - 17786005 on NCBI Build GRCm38
          Downstream cysteine-rich perinuclear theca 12 Cypt12 1700001F04Rik,
          AV209098
          148.4kb Sense Chr. 3 - 17948444 - 17948952 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000112.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447980 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177530 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151414 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172898 AK080704.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172898 AK163433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172898 BE647934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172898 AK011787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172898 NR_033490.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04836118 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151414 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177530 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04836109 - -
          Whole Mouse Genome G4122A A_51_P403704 AK163433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403704 AK011787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403704 NR_033490.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111686 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123702 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177530 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03144260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21968104 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300000489.mm.1 TC0300000112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000489.mm.1 NR_033490.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000489.mm.1 NR_033490.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000486.mm.1 TC0300000112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000486.mm.1 NR_033490.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000486.mm.1 NR_033490.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000490.mm.1 TC0300000112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300000490.mm.1 NR_033490.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300000490.mm.1 NR_033490.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 697306 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0