Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292449_m1
          See other CCR10 GE Assays ›
          Gene Symbol:
          Ccr10
          Gene Name:
          chemokine (C-C motif) receptor 10
          Gene Aliases:
          Cmkbr9, Gpr2
          Chromosome Location:
          Chr.11: 101172922 - 101175695 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292449_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccr10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007721.4 NP_031747.2 1 - 2 44 72
          GenBank mRNA AK090044.1 - 1 - 2 47 72
          BC117043.1 - 1 - 2 47 40126362 72
          BC120542.1 - 1 - 2 47 40129465 72

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK090044.1, BC117043.1, BC120542.1, NM_007721.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccr10

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          immune response
          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          chemokine-mediated signaling pathway
          signal transducer activity
          G-protein coupled receptor activity
          chemokine receptor activity
          protein binding
          C-C chemokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072389 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072388 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072390 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105878 AK090044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105878 NM_007721.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105878 AF215983.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403347 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116148 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105878 AK090044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105878 NM_007721.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105878 AF215983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395877 AK154468.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395877 BC120542.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395877 AK090044.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395877 BC117043.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395877 NM_007721.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395877 AK154468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395877 BC120542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395877 AK090044.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395877 BC117043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395877 NM_007721.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127520 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09710537 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09710538 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430353 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09430352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09479891 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072389 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072388 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28072390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28072374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09710539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09314690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09479891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28072379 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391376 - -
          Affymetrix HT MG-430 PM Array Plate 1421420_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391373 - -
          Mouse Genome 430 2.0 Array 1421420_at - -
          Mouse Genome 430 2.0 Array 1421420_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12777_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12777_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100035086.mm.1 NM_007721.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035086.mm.1 NM_007721.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035086.mm.1 TC1100003796.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035085.mm.1 NM_007721.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035085.mm.1 NM_007721.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035085.mm.1 TC1100003796.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035087.mm.1 NM_007721.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035087.mm.1 NM_007721.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035087.mm.1 TC1100003796.mm.1 MTA TC
          Mouse Expression Set 430 1421420_at - -
          Mouse Expression Set 430 1421420_PM_at - -
          Mouse Genome 430A 2.0 Array 1421420_PM_at - -
          Mouse Genome 430A 2.0 Array 1421420_at - -
          Affymetrix HT Mouse Genome 430A Array 1421420_at - -
          Affymetrix HT Mouse Genome 430A Array 1421420_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 536328 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4050097 - -
          Illumina mouse-6 v1.0 expression beadchip 4050097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230042 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230042 - -
          mouse-6 v1.1 (Illumina) scl39541.2.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39541.2.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230042 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix