Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292788_m1
          See other 4930558K02RIK GE Assays ›
          Gene Symbol:
          4930558K02Rik
          Gene Name:
          RIKEN cDNA 4930558K02 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 161942089 - 161982371 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292788_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930558K02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204904.1 NP_001191833.1 6 - 7 531 72
          NM_001204905.1 NP_001191834.1 6 - 7 564 72
          GenBank mRNA AK016179.1 - 6 - 7 567 72
          CF105154.1 - 6 - 7 531 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SUN domain containing ossification factor Suco 4732491B11,
          AI848100,
          AU016931,
          Opt
          -65.4kb Sense Chr. 1 - 161816112 - 161876661 on NCBI Build GRCm38
          Downstream phosphatidylinositol glycan anchor biosynthesis,
          class C
          Pigc 3110030E07Rik,
          AW212108
          19.5kb Anti-sense Chr. 1 - 161969188 - 161973435 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003381.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2376547 NM_001204904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2376547 NM_001204905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2376547 AK016179.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29460 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131150 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2376547 NM_001204904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2376547 NM_001204905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2376547 AK016179.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00234577 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131150 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04331983 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00234565 - -
          Whole Mouse Genome G4122A A_52_P445547 NM_001204904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P445547 NM_001204905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P445547 AK016179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P445547 CF105154.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20774668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00268817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00612193 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10359474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10359476 - -
          Affymetrix HT MG-430 PM Array Plate 1431636_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10359473 - -
          Mouse Genome 430 2.0 Array 1431636_at - -
          Mouse Genome 430 2.0 Array 1431636_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100027686.mm.1 TC0100003381.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100027686.mm.1 NM_001204904.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100027686.mm.1 NM_001204904.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100027686.mm.1 NM_001204905.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100027686.mm.1 NM_001204905.1 Refseq
          Mouse Expression Set 430 1431636_PM_at - -
          Mouse Expression Set 430 1431636_at - -
          Affymetrix HT Mouse Genome 430B Array 1431636_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431636_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309885 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix