Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295331_g1
          See other PPOX GE Assays ›
          Gene Symbol:
          Ppox
          Gene Name:
          protoporphyrinogen oxidase
          Gene Aliases:
          Ppo
          Chromosome Location:
          Chr.1: 171276991 - 171281203 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295331_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppox
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008911.2 NP_032937.1 12 - 13 1613 88
          GenBank mRNA AK134341.1 - 3142 88
          AK142632.1 - 12 - 13 1565 88
          BC002047.1 - 12 - 13 1568 3484079 88
          D45185.1 - 12 - 13 88
          U25114.1 - 12 - 13 1588 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003536.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppox

          Gene Ontology Categories:

          Function(s) Process(es)

          porphyrin-containing compound biosynthetic process
          heme biosynthetic process
          protoporphyrinogen IX metabolic process
          oxygen-dependent protoporphyrinogen oxidase activity
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P310949 D45185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P310949 U25114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P310949 BC002047.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P310949 D45185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P310949 U25114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P310949 BC002047.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470184 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P310949 D45185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P310949 U25114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P310949 BC002047.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P310949 D45185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P310949 U25114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P310949 BC002047.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P310949 D45185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P310949 U25114.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P310949 BC002047.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P310949 D45185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P310949 U25114.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P310949 BC002047.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827080 - -
          Whole Mouse Genome G4122A A_51_P310949 D45185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P310949 U25114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P310949 BC002047.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P310949 D45185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P310949 U25114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P310949 BC002047.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131570 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128645 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00481670 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00481633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00481630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00481666 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00481626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00481634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00786864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00786863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00481633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360102 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360103 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360091 - -
          Affymetrix HT MG-430 PM Array Plate 1441862_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416618_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360090 - -
          Mouse Genome 430 2.0 Array 1416618_at - -
          Mouse Genome 430 2.0 Array 1416618_PM_at - -
          Mouse Genome 430 2.0 Array 1448065_PM_at - -
          Mouse Genome 430 2.0 Array 1441862_at - -
          Mouse Genome 430 2.0 Array 1441862_PM_at - -
          Mouse Genome 430 2.0 Array 1448065_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19044_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19044_at - -
          Affymetrix Murine Genome 6500A Array Msa17350_at - -
          Affymetrix Mouse Clariom S Array PSR0100028942.mm.1 TC0100003536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028942.mm.1 NM_008911.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028942.mm.1 NM_008911.2 Refseq
          Mouse Expression Set 430 1416618_at - -
          Mouse Expression Set 430 1416618_PM_at - -
          Mouse Expression Set 430 1441862_at - -
          Mouse Expression Set 430 1441862_PM_at - -
          Murine Genome U74v2 Set 100429_at - -
          Murine Genome U74v2 Set 100429_PM_at - -
          Mouse Genome 430A 2.0 Array 1416618_PM_at - -
          Mouse Genome 430A 2.0 Array 1416618_at - -
          Affymetrix HT Mouse Genome 430A Array 1416618_at - -
          Affymetrix HT Mouse Genome 430A Array 1416618_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441862_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441862_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 820089 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850102 - -
          Illumina mouse-6 v1.0 expression beadchip 1240672 - -
          Illumina mouse-6 v1.0 expression beadchip 3850102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706938 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618316 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618316 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706938 - -
          mouse-6 v1.1 (Illumina) scl15943.8.1_52-S - -
          mouse-6 v1.1 (Illumina) scl000961.1_62-S - -
          Illumina Sentrix mouse-6.v1 bead array 1240672 - -
          Illumina Sentrix mouse-6.v1 bead array 3850102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15943.8.1_52-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000961.1_62-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850102 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2826816 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706938 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706938 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2826816 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl15943.8.1_52-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0