Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200011795.mm.1 |
TC1200001715.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011795.mm.1 |
NM_001172098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011795.mm.1 |
NM_001172098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011795.mm.1 |
NM_052973.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011795.mm.1 |
NM_052973.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011796.mm.1 |
TC1200001715.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011796.mm.1 |
NM_001172098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011796.mm.1 |
NM_001172098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011796.mm.1 |
NM_052973.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011796.mm.1 |
NM_052973.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011794.mm.1 |
TC1200001715.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011794.mm.1 |
NM_001172098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011794.mm.1 |
NM_001172098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011794.mm.1 |
NM_052973.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011794.mm.1 |
NM_052973.2 |
Refseq |