Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01296073_m1
          See other LY9 GE Assays ›
          Gene Symbol:
          Ly9
          Gene Name:
          lymphocyte antigen 9
          Gene Aliases:
          AI893573, CD229, Lgp100, Ly-9, SLAMF3, T100
          Chromosome Location:
          Chr.1: 171588613 - 171607410 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01296073_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277968.1 NP_001264897.1 8 - 9 1659 148
          NM_008534.3 NP_032560.2 9 - 10 1989 148
          NR_102726.1 9 - 10 2005 148
          GenBank mRNA AF244130.1 - 9 - 10 1989 148
          AF244131.1 - 9 - 10 1989 148
          AK088815.1 - 8 - 9 1608 148
          AK134106.1 - 9 - 10 2006 148
          AK150121.1 - 9 - 10 1915 148
          BC055380.1 - 9 - 10 1889 148
          BC066212.1 - 9 - 10 1877 148
          BC095921.1 - 9 - 10 1912 4935844 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003549.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly9

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          signaling receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497171 M84412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497171 M84412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989976 BC095921.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989976 M84412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989976 BC055380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989976 BC066212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989976 AK134106.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30028 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129385 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126274 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989976 BC095921.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989976 M84412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989976 BC055380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989976 BC066212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989976 AK134106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497171 M84412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497171 M84412.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497171 M84412.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497171 M84412.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20828751 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00251173 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04354752 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129385 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126274 - -
          Whole Mouse Genome G4122A A_51_P497171 M84412.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497171 M84412.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121264 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111928 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129385 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01096088 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00037770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00871244 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00871246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20828574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00266023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01096089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00266019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01128103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00266013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00266021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01096088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00684765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01128102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01128100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00236118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00181258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01128093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00322476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01128099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00205546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00266020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00037770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00684761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20828766 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360165 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360164 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360171 - -
          Affymetrix HT MG-430 PM Array Plate 1449156_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360158 - -
          Mouse Genome 430 2.0 Array 1449156_at - -
          Mouse Genome 430 2.0 Array 1449156_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17085_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17085_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100029129.mm.1 TC0100003549.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029129.mm.1 NM_001277968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029129.mm.1 NM_008534.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029129.mm.1 NR_102726.1 Refseq
          Mouse Expression Set 430 1449156_PM_at - -
          Mouse Expression Set 430 1449156_at - -
          Murine Genome U74v2 Set 93194_at - -
          Murine Genome U74v2 Set 93194_PM_at - -
          Mouse Genome 430A 2.0 Array 1449156_at - -
          Mouse Genome 430A 2.0 Array 1449156_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449156_at - -
          Affymetrix HT Mouse Genome 430A Array 1449156_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4070292 - -
          Illumina mouse-6 v1.0 expression beadchip 4070292 - -
          Illumina mouse-6 v1.0 expression beadchip 103390215 - -
          Illumina mouse-6 v1.0 expression beadchip 3360315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2764169 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231309 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231309 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213156 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2764169 - -
          mouse-6 v1.1 (Illumina) scl15933.11.1_5-S - -
          mouse-6 v1.1 (Illumina) scl000777.1_104-S - -
          Illumina Sentrix mouse-6.v1 bead array 103390215 - -
          Illumina Sentrix mouse-6.v1 bead array 4070292 - -
          Illumina Sentrix mouse-6.v1 bead array 3360315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15933.11.1_5-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000777.1_104-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360315 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070292 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103390215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213596 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231309 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213156 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213596 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000777.1_104-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0