Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01297742_m1
          See other PSEN1 GE Assays ›
          Gene Symbol:
          Psen1
          Gene Name:
          presenilin 1
          Gene Aliases:
          Ad3h, PS-1, PS1, S182
          Chromosome Location:
          Chr.12: 83688146 - 83738446 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01297742_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psen1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008943.2 NP_032969.1 7 - 8 1261 64
          GenBank mRNA AF149111.1 - 5 - 6 773 64
          AK134232.1 - 7 - 8 944 64
          AK134615.1 - 7 - 8 983 64
          AK148101.1 - 7 - 8 982 64
          AK165984.1 - 7 - 8 982 64
          AK169963.1 - 3 - 4 388 64
          AK205380.1 - 1 - 2 32 64
          BC030409.1 - 7 - 8 936 5133302 64
          BC071233.1 - 7 - 8 1000 3673609 64
          L42177.1 - 7 - 8 957 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000792.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psen1

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          negative regulation of transcription by RNA polymerase II
          blood vessel development
          cell fate specification
          somitogenesis
          neuron migration
          positive regulation of receptor recycling
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart looping
          hematopoietic progenitor cell differentiation
          T cell activation involved in immune response
          myeloid leukocyte differentiation
          negative regulation of protein kinase activity
          protein glycosylation
          proteolysis
          membrane protein ectodomain proteolysis
          calcium ion transport
          mitochondrial transport
          cellular calcium ion homeostasis
          autophagy
          apoptotic process
          cellular response to DNA damage stimulus
          response to oxidative stress
          cell adhesion
          negative regulation of epidermal growth factor-activated receptor activity
          regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          Notch receptor processing
          brain development
          heart development
          learning or memory
          memory
          embryo development
          post-embryonic development
          protein transport
          L-glutamate transmembrane transport
          choline transport
          synaptic vesicle targeting
          protein processing
          cerebral cortex cell migration
          Cajal-Retzius cell differentiation
          dorsal/ventral neural tube patterning
          cerebral cortex development
          neurogenesis
          neuron differentiation
          embryonic limb morphogenesis
          forebrain development
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          endoplasmic reticulum calcium ion homeostasis
          amyloid-beta formation
          segmentation
          intracellular signal transduction
          locomotion
          regulation of phosphorylation
          anagen
          amyloid precursor protein catabolic process
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          regulation of protein binding
          positive regulation of MAP kinase activity
          negative regulation of neuron apoptotic process
          skin morphogenesis
          cellular protein metabolic process
          positive regulation of protein kinase activity
          positive regulation of transcription, DNA-templated
          regulation of synaptic plasticity
          thymus development
          neuron development
          skeletal system morphogenesis
          brain morphogenesis
          amyloid-beta metabolic process
          epithelial cell proliferation
          negative regulation of axonogenesis
          positive regulation of coagulation
          T cell receptor signaling pathway
          neuron apoptotic process
          negative regulation of ubiquitin-protein transferase activity
          smooth endoplasmic reticulum calcium ion homeostasis
          protein maturation
          regulation of synaptic transmission, glutamatergic
          canonical Wnt signaling pathway
          regulation of resting membrane potential
          positive regulation of dendritic spine development
          calcium ion transmembrane transport
          cell-cell adhesion
          negative regulation of ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic signaling pathway
          endopeptidase activity
          aspartic-type endopeptidase activity
          calcium channel activity
          protein binding
          beta-catenin binding
          peptidase activity
          hydrolase activity
          PDZ domain binding
          aspartic endopeptidase activity, intramembrane cleaving
          cadherin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200005622.mm.1 TC1200000792.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200005622.mm.1 NM_008943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200005622.mm.1 NM_008943.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0