Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029447.2 | NP_083723.1 | 10 - 11 | 1870 | 94 | |
GenBank mRNA | AK079090.1 | - | 10 - 11 | 1871 | 94 | |
AK138399.1 | - | 10 - 11 | 1843 | 94 | ||
AK146364.1 | - | 10 - 11 | 1819 | 94 | ||
AK170508.1 | - | 10 - 11 | 1871 | 94 | ||
AK171432.1 | - | 10 - 11 | 1871 | 94 | ||
BC016224.1 | - | 10 - 11 | 1843 | 4486941 | 94 | |
BC025520.1 | - | 3 - 4 | 729 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |