Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014392.mm.1 |
TC1700001626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014392.mm.1 |
NM_025718.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014392.mm.1 |
NM_025718.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014389.mm.1 |
TC1700001626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014389.mm.1 |
NM_025718.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014389.mm.1 |
NM_025718.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014390.mm.1 |
TC1700001626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014390.mm.1 |
NM_025718.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014390.mm.1 |
NM_025718.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014395.mm.1 |
TC1700001626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014395.mm.1 |
NM_025718.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014395.mm.1 |
NM_025718.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014393.mm.1 |
TC1700001626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014393.mm.1 |
NM_025718.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014393.mm.1 |
NM_025718.3 |
Refseq |