Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01300355_g1
          See other ACVRL1 GE Assays ›
          Gene Symbol:
          Acvrl1
          Gene Name:
          activin A receptor, type II-like 1
          Gene Aliases:
          AI115505, AI427544, Acvrlk1, Alk1
          Chromosome Location:
          Chr.15: 101128522 - 101145336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01300355_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acvrl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277255.1 NP_001264184.1 8 - 9 1060 82
          NM_001277257.1 NP_001264186.1 7 - 8 999 82
          NM_001277258.1 NP_001264187.1 7 - 8 968 82
          NM_001277259.1 NP_001264188.1 8 - 9 1055 82
          NM_009612.3 NP_033742.2 7 - 8 1037 82
          GenBank mRNA AK134712.1 - 5 - 6 794 82
          AK160915.1 - 8 - 9 1028 82
          AK170870.1 - 5 - 6 793 82
          BC014291.1 - 6 - 7 956 3982004 82
          BC015083.1 - 6 - 7 918 3980151 82
          CT010330.1 - 5 - 6 765 82
          L48015.1 - 7 - 8 1039 82
          Z31664.1 - 7 - 8 1005 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001061.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acvrl1

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          in utero embryonic development
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          blood vessel remodeling
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of cell adhesion
          signal transduction
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          blood circulation
          regulation of blood pressure
          negative regulation of cell population proliferation
          negative regulation of endothelial cell migration
          positive regulation of pathway-restricted SMAD protein phosphorylation
          phosphorylation
          negative regulation of cell growth
          negative regulation of cell migration
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          activin receptor signaling pathway
          wound healing, spreading of epidermal cells
          intracellular signal transduction
          endothelial cell activation
          negative regulation of blood vessel endothelial cell migration
          negative regulation of endothelial cell differentiation
          positive regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          blood vessel morphogenesis
          protein heterooligomerization
          negative regulation of focal adhesion assembly
          lymphatic endothelial cell differentiation
          artery development
          venous blood vessel development
          endothelial tube morphogenesis
          retina vasculature development in camera-type eye
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          negative regulation of DNA biosynthetic process
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          kinase activity
          activin receptor activity, type I
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500008447.mm.1 TC1500001061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008447.mm.1 NM_001277255.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008447.mm.1 NM_001277257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008447.mm.1 NM_001277258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008447.mm.1 NM_001277259.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008447.mm.1 NM_009612.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008446.mm.1 TC1500001061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008446.mm.1 NM_001277255.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008446.mm.1 NM_001277257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008446.mm.1 NM_001277258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008446.mm.1 NM_001277259.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008446.mm.1 NM_009612.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0