Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01301135_g1
          See other JMJD8 GE Assays ›
          Gene Symbol:
          Jmjd8
          Gene Name:
          jumonji domain containing 8
          Gene Aliases:
          2610003J06Rik
          Chromosome Location:
          Chr.17: 25829043 - 25831843 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01301135_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jmjd8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028101.4 NP_082377.2 8 - 9 758 63
          GenBank mRNA AK011308.1 - 8 - 9 755 63
          AK148079.1 - 8 - 9 736 63
          AK155810.1 - 8 - 9 760 63
          AK172073.1 - 8 - 9 755 63
          BC010800.1 - 8 - 9 720 4235072 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000391.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jmjd8

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434776 AK011308.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434776 AK011308.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55902 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175261 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434776 AK011308.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434776 AK011308.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31106568 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175261 - -
          Whole Mouse Genome G4122A A_51_P434776 AK011308.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434776 AK011308.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127503 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13128245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13128243 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13128248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31106599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13351714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13128247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31106550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442909 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442913 - -
          Affymetrix HT MG-430 PM Array Plate 1424743_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442904 - -
          Affymetrix Mouse Gene 1.1 ST Array 10449051 - -
          Mouse Genome 430 2.0 Array 1424743_PM_at - -
          Mouse Genome 430 2.0 Array 1424743_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72106_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72106_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700003321.mm.1 TC1700000391.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003321.mm.1 NM_028101.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003321.mm.1 NM_028101.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003320.mm.1 TC1700000391.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003320.mm.1 NM_028101.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003320.mm.1 NM_028101.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003318.mm.1 TC1700000391.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003318.mm.1 NM_028101.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003318.mm.1 NM_028101.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003322.mm.1 TC1700000391.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003322.mm.1 NM_028101.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003322.mm.1 NM_028101.4 Refseq
          Mouse Expression Set 430 1424743_at - -
          Mouse Expression Set 430 1424743_PM_at - -
          Murine Genome U74v2 Set 94520_at - -
          Murine Genome U74v2 Set 94520_PM_at - -
          Mouse Genome 430A 2.0 Array 1424743_at - -
          Mouse Genome 430A 2.0 Array 1424743_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424743_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424743_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 485403 - -
          ABI Mouse Genome Survey Microarray 759175 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450575 - -
          Illumina mouse-6 v1.0 expression beadchip 6450575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450575 - -
          mouse-6 v1.1 (Illumina) scl072106.9_198-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl072106.9_198-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2896492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896492 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl072106.9_198-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0