Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303079_g1
          See other ZFP207 GE Assays ›
          Gene Symbol:
          Zfp207
          Gene Name:
          zinc finger protein 207
          Gene Aliases:
          8430401D15Rik, BuGZ, Zep, Znf207
          Chromosome Location:
          Chr.11: 80383279 - 80405992 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303079_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130169.1 NP_001123641.1 7 - 8 849 90
          NM_001130170.1 NP_001123642.1 6 - 7 801 90
          NM_001130171.1 NP_001123643.1 7 - 8 849 90
          NR_045038.1 7 - 8 849 90
          GenBank mRNA AB032196.1 - 7 - 8 730 90
          AK033497.1 - 7 - 8 798 90
          AK087885.1 - 7 - 8 849 90
          AK140139.1 - 6 - 7 785 90
          AK146787.1 - 7 - 8 812 90
          AK159683.1 - 7 - 8 800 90
          BC003715.1 - 7 - 8 790 3601357 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001204.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          mitotic cell cycle
          cell cycle
          chromosome segregation
          mitotic spindle assembly checkpoint signaling
          attachment of spindle microtubules to kinetochore
          protein stabilization
          cell division
          regulation of chromosome segregation
          RNA binding
          microtubule binding
          heparin binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 TC1100001204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_001130169.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_001130169.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_001130170.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_001130170.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_001130171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_001130171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_011751.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NM_011751.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NR_045038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011268.mm.1 NR_045038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 TC1100001204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_001130169.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_001130169.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_001130170.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_001130170.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_001130171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_001130171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_011751.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NM_011751.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NR_045038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011269.mm.1 NR_045038.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0