Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100032077.mm.1 |
TC1100003505.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100032077.mm.1 |
NM_001042541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032077.mm.1 |
NM_001042541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032077.mm.1 |
NM_009648.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032077.mm.1 |
NM_009648.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032077.mm.1 |
NR_104310.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032079.mm.1 |
TC1100003505.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100032079.mm.1 |
NM_001042541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032079.mm.1 |
NM_001042541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032079.mm.1 |
NM_009648.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032079.mm.1 |
NM_009648.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032079.mm.1 |
NR_104310.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032078.mm.1 |
TC1100003505.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100032078.mm.1 |
NM_001042541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032078.mm.1 |
NM_001042541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032078.mm.1 |
NM_009648.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032078.mm.1 |
NM_009648.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100032078.mm.1 |
NR_104310.1 |
Refseq |