Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159509.1 | NP_001152981.1 | 5 - 6 | 823 | 84 | |
NM_001159510.1 | NP_001152982.1 | 5 - 6 | 774 | 84 | ||
NM_011862.3 | NP_035992.1 | 5 - 6 | 862 | 84 | ||
GenBank mRNA | AF128535.1 | - | 5 - 6 | 852 | 84 | |
AK154785.1 | - | 5 - 6 | 749 | 84 | ||
AK170299.1 | - | 5 - 6 | 821 | 84 | ||
CA560112.1 | - | 3 - 4 | 507 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() |