Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016450.mm.1 |
TC1500002048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016450.mm.1 |
NM_001081030.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016450.mm.1 |
NM_001081030.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016450.mm.1 |
NM_001170561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016450.mm.1 |
NM_001170561.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016449.mm.1 |
TC1500002048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016449.mm.1 |
NM_001081030.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016449.mm.1 |
NM_001081030.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016449.mm.1 |
NM_001170561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016449.mm.1 |
NM_001170561.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016448.mm.1 |
TC1500002048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016448.mm.1 |
NM_001081030.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016448.mm.1 |
NM_001081030.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016448.mm.1 |
NM_001170561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016448.mm.1 |
NM_001170561.1 |
Refseq |