Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303683_m1
          See other 9430014N10RIK GE Assays ›
          Gene Symbol:
          9430014N10Rik
          Gene Name:
          RIKEN cDNA 9430014N10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 93904856 - 93927111 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303683_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430014N10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045737.1 2 - 3 321 132
          GenBank mRNA AK020418.1 - 2 - 3 322 132
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY721595.1 - 2 - 3 322 Chr. 15 - 93905965 - 93927111 [-] on Build GRCm38 132

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          30339
          Gm30339 -163.9kb Anti-sense Chr. 15 - 93706120 - 93741854 on NCBI Build GRCm38
          Downstream predicted gene,
          30564
          Gm30564 38.4kb Sense Chr. 15 - 93956424 - 93964708 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002105.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2389714 AK020418.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136862 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08465841 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130949 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08465841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12403970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12326825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12037024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12132132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12278905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12148509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12403975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12087087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12148513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12149420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12326815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12054470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12217171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12440507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30449901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432258_PM_at - -
          Mouse Genome 430 2.0 Array 1432258_at - -
          Mouse Genome 430 2.0 Array 1432258_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500016899.mm.1 TC1500002105.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016899.mm.1 NR_045737.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016899.mm.1 NR_045737.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016900.mm.1 TC1500002105.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016900.mm.1 NR_045737.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016900.mm.1 NR_045737.1 Refseq
          Mouse Expression Set 430 1432258_at - -
          Mouse Expression Set 430 1432258_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432258_at - -
          Affymetrix HT Mouse Genome 430B Array 1432258_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 497896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0