Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01304060_m1
          See other 4930556M19RIK GE Assays ›
          Gene Symbol:
          4930556M19Rik
          Gene Name:
          RIKEN cDNA 4930556M19 gene
          Gene Aliases:
          1700009D01Rik, C130022M03Rik
          Chromosome Location:
          Chr.15: 10714836 - 10790123 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01304060_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930556M19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045063.1 4 - 5 496 83
          NR_045064.1 4 - 5 435 83
          GenBank mRNA AI552009.1 - 2 - 3 170 83
          AK016150.1 - 4 - 5 437 83
          AK030258.1 - 4 - 5 497 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB617275.1 - 4 - 5 497 Chr. 15 - 10714836 - 10769215 [+] on Build GRCm38 83
          GenBankmRNA BY715753.1 - 4 - 5 437 Chr. 15 - 10714839 - 10769333 [+] on Build GRCm38 83
          GenBankmRNA AA072067.1 - 2 - 3 169 Chr. 15 - 10767490 - 10769303 [+] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream retinoic acid induced 14 Rai14 1700008J19Rik,
          1700020L11Rik,
          Ankycorbin,
          Norpeg,
          mKIAA1334
          -48.8kb Anti-sense Chr. 15 - 10568971 - 10719267 on NCBI Build GRCm38
          Downstream predicted gene,
          19276
          Gm19276 59.7kb Sense Chr. 15 - 10829051 - 10832713 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000084.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738829 M69200.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738829 NM_009377.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136224 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116837 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P182391 AK047371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P182391 AK047938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P182391 AK005793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P182391 AK047371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P182391 AK047938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P182391 AK005793.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02972977 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08283221 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08283303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08283238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02972989 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08283353 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02973053 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116837 - -
          Whole Mouse Genome G4122A A_52_P241872 AK030258.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02972989 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12125139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12224943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12322189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12431117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12270140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12068127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11884335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12020127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12125134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11884340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12125150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12076678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12322186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12271926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11856708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11884330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12020131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12076679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11914048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12271925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12101886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12020203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12431124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12217358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12076682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12020125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30012375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500000793.mm.1 TC1500000084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000793.mm.1 NR_045063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000793.mm.1 NR_045063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000793.mm.1 NR_045064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000793.mm.1 NR_045064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000807.mm.1 TC1500000084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000807.mm.1 NR_045063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000807.mm.1 NR_045063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000807.mm.1 NR_045064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000807.mm.1 NR_045064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000809.mm.1 TC1500000084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000809.mm.1 NR_045063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000809.mm.1 NR_045063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000809.mm.1 NR_045064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000809.mm.1 NR_045064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000808.mm.1 TC1500000084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000808.mm.1 NR_045063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000808.mm.1 NR_045063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000808.mm.1 NR_045064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000808.mm.1 NR_045064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000798.mm.1 TC1500000084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000798.mm.1 NR_045063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000798.mm.1 NR_045063.1 Refseq
          Mouse Expression Set 430 1447451_PM_at - -
          Mouse Expression Set 430 1447451_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103840148 - -
          MouseWG-6 v2 Expression BeadChip 102320706 - -
          Illumina mouse-6 v1.0 expression beadchip 102320706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2459394 - -
          mouse-6 v1.1 (Illumina) scl0075259.1_33-S - -
          Illumina Sentrix mouse-6.v1 bead array 102320706 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102320706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0