Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01304172_m1
          See other RNF135 GE Assays ›
          Gene Symbol:
          Rnf135
          Gene Name:
          ring finger protein 135
          Gene Aliases:
          0610037N03Rik, 2410006N06Rik
          Chromosome Location:
          Chr.11: 80183825 - 80199754 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01304172_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf135
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028019.3 NP_082295.1 4 - 5 767 59
          GenBank mRNA AK010429.1 - 4 - 5 769 59
          AK150921.1 - 4 - 5 768 59
          AK152096.1 - 4 - 5 760 59
          AK185113.1 - 3 - 4 357 59
          AK206997.1 - 3 - 4 357 59
          BC096385.1 - 4 - 5 761 30744704 59
          BC132307.1 - 4 - 5 763 40130584 59
          BC138303.1 - 4 - 5 763 8861322 59
          BC145236.1 - 3 - 4 599 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001197.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf135

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          protein ubiquitination
          positive regulation of interferon-beta production
          innate immune response
          regulation of innate immune response
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          ribonucleoprotein complex binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK182968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 BC025209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK152096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK002781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK010429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK182968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 BC025209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK010429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK152096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P367285 AK002781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084112 BC132307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084112 BC138303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084112 BC145236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084112 BC025209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084112 BC096385.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111577 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129804 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152177 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084112 BC132307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084112 BC138303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084112 BC145236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084112 BC025209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084112 BC096385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK182968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 BC025209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK152096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK002781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK010429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK182968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 BC025209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK152096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK002781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P367285 AK010429.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27952998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27953004 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129804 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07412861 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02373789 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152177 - -
          Whole Mouse Genome G4122A A_51_P367285 AK182968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 BC025209.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK152096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK002781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK010429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK182968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 BC025209.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK152096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK002781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367285 AK010429.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111368 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152177 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129804 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09485925 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09485929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09485921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27953115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09276648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09485926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09304894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27953029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09093329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379407 - -
          Affymetrix HT MG-430 PM Array Plate 1452289_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379401 - -
          Mouse Genome 430 2.0 Array 1452289_a_at - -
          Mouse Genome 430 2.0 Array 1452289_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71956_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71956_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100011179.mm.1 TC1100001197.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011179.mm.1 NM_028019.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011179.mm.1 NM_028019.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011180.mm.1 TC1100001197.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011180.mm.1 NM_028019.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011180.mm.1 NM_028019.3 Refseq
          Mouse Expression Set 430 1452289_PM_a_at - -
          Mouse Expression Set 430 1452289_a_at - -
          Mouse Genome 430A 2.0 Array 1452289_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1452289_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452289_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452289_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 925331 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6180390 - -
          Illumina mouse-6 v1.0 expression beadchip 1660685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180390 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655034 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259341 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655034 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259341 - -
          mouse-6 v1.1 (Illumina) scl0001435.1_11-S - -
          mouse-6 v1.1 (Illumina) scl41177.6_160-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180390 - -
          Illumina Sentrix mouse-6.v1 bead array 1660685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41177.6_160-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001435.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2820379 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2820383 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655034 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2820379 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2820383 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655034 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001435.1_11-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix