Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810021B22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040417.1 | 2 - 3 | 366 | 88 | ||
GenBank mRNA | AK007571.1 | - | 2 - 3 | 367 | 88 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pannexin 2 | Panx2 | Anti-sense | Chr. 15 - 89059726 - 89073571 on NCBI Build GRCm38 | ||
Downstream | TraB domain containing | Trabd |
5730502D15Rik, AL023039 |
1.9kb | Anti-sense | Chr. 15 - 89076050 - 89087078 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |