Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300020169.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020169.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020169.mm.1 |
NM_010763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020175.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020175.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020175.mm.1 |
NM_010763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020173.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020173.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020173.mm.1 |
NM_010763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020176.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020176.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020176.mm.1 |
NM_010763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020170.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020170.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020170.mm.1 |
NM_010763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020177.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020177.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020177.mm.1 |
NM_010763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020174.mm.1 |
TC0300002583.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020174.mm.1 |
NM_010763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020174.mm.1 |
NM_010763.2 |
Refseq |