Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310827_m1
          See other FAM228A GE Assays ›
          Gene Symbol:
          Fam228a
          Gene Name:
          family with sequence similarity 228, member A
          Gene Aliases:
          4930417G10Rik
          Chromosome Location:
          Chr.12: 4697272 - 4738468 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310827_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam228a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029107.2 NP_083383.1 7 - 8 931 70
          GenBank mRNA AK029492.1 - 7 - 8 932 70
          AK133272.1 - 7 - 8 969 70
          BC138846.1 - 7 - 8 866 8861868 70
          BC138853.1 - 7 - 8 774 8861875 70

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam228a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122433 AK029492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122433 AK133272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122433 NM_029107.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116402 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108386 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170115 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122433 AK029492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122433 AK133272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122433 NM_029107.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202189 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170115 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108386 - -
          Whole Mouse Genome G4122A A_52_P394506 BC138846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394506 BC138853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394506 AK029492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394506 NM_029107.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P394506 AK015158.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122433 AK029492.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122433 AK133272.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122433 NM_029107.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163674 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09857660 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09857658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10087678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10083617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10400424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10024810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09804494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10231313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10132497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10378675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09804462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09804488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10024818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10231312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10083628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10231314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09804487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10289985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202211 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10399297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399294 - -
          Affymetrix HT MG-430 PM Array Plate 1432283_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10399290 - -
          Mouse Genome 430 2.0 Array 1432283_at - -
          Mouse Genome 430 2.0 Array 1432283_PM_at - -
          Mouse Expression Set 430 1432283_at - -
          Mouse Expression Set 430 1432283_PM_at - -
          Murine Genome U74v2 Set 167770_PM_at - -
          Murine Genome U74v2 Set 167770_at - -
          Affymetrix HT Mouse Genome 430B Array 1432283_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432283_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632144 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106040452 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222994 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255040 - -
          Illumina Sentrix mouse-6.v1 bead array 106040452 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106040452 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255040 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix