Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310894_m1
          See other FAM228B GE Assays ›
          Gene Symbol:
          Fam228b
          Gene Name:
          family with sequence similarity 228, member B
          Gene Aliases:
          A830093I24Rik
          Chromosome Location:
          Chr.12: 4738397 - 4769267 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310894_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam228b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175431.4 NP_780640.2 3 - 4 563 144
          GenBank mRNA AK044133.1 - 3 - 4 564 144
          AK050447.1 - 3 - 4 529 144
          BC100429.1 - 3 - 4 342 30917787 144
          BC144941.1 - 3 - 4 294 144
          BC145648.1 - 3 - 4 294 40130951 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001364.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam228b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202317 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202318 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 BC145648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 AK050447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 BC144941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 NM_175431.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 BC145648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 AK050447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 BC144941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P82617 NM_175431.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118709 BC145648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118709 AK050447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118709 BC100429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118709 BC144941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118709 NM_175431.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081697 BC145648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081697 AK050447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081697 BC100429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081697 BC144941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081697 NM_175431.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33704 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180577 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141162 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081697 BC145648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081697 AK050447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081697 BC100429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081697 BC144941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081697 NM_175431.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118709 BC145648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118709 AK050447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118709 BC100429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118709 BC144941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118709 NM_175431.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 BC145648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 AK050447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 BC144941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 NM_175431.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 BC145648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 AK050447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 BC144941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P82617 NM_175431.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02450575 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180577 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07524322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141162 - -
          Whole Mouse Genome G4122A A_52_P82617 BC145648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 AK050447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 BC144941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 NM_175431.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 BC145648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 AK050447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 BC144941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P82617 NM_175431.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148717 BC145648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148717 AK050447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148717 BC100429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148717 BC144941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148717 NM_175431.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108675 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180577 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02450575 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141162 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09906321 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09906322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202318 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09915362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10052354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10399306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399303 - -
          Affymetrix Mouse Gene 1.1 ST Array 10399299 - -
          Affymetrix Mouse Clariom S Array PSR1200009770.mm.1 TC1200001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009770.mm.1 NM_175431.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009770.mm.1 NM_175431.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009767.mm.1 TC1200001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009767.mm.1 NM_175431.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009767.mm.1 NM_175431.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009768.mm.1 TC1200001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009768.mm.1 NM_175431.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009768.mm.1 NM_175431.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009776.mm.1 TC1200001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009776.mm.1 NM_175431.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009776.mm.1 NM_175431.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009775.mm.1 TC1200001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009775.mm.1 NM_175431.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009775.mm.1 NM_175431.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009773.mm.1 TC1200001364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009773.mm.1 NM_175431.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009773.mm.1 NM_175431.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 805887 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100360592 - -
          Illumina mouse-6 v1.0 expression beadchip 4150605 - -
          Illumina mouse-6 v1.0 expression beadchip 100360592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150605 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255757 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221166 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255757 - -
          mouse-6 v1.1 (Illumina) scl0207921.1_329-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150605 - -
          Illumina Sentrix mouse-6.v1 bead array 100360592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0207921.1_329-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100360592 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255757 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221166 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix