Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01314897_g1
          See other CMTM5 GE Assays ›
          Gene Symbol:
          Cmtm5
          Gene Name:
          CKLF-like MARVEL transmembrane domain containing 5
          Gene Aliases:
          1500005P16Rik, 2900052H21Rik, Cklfsf5
          Chromosome Location:
          Chr.14: 54936470 - 54939277 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01314897_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cmtm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026066.2 NP_080342.1 4 - 5 644 79
          NM_029390.1 NP_083666.1 3 - 4 464 79
          GenBank mRNA AK005168.1 - 3 - 4 507 79
          AK013666.1 - 4 - 5 644 79
          AY241867.1 - 4 - 5 634 79
          BC049665.1 - 4 - 5 631 6533928 79
          DQ115983.1 - 3 - 4 464 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000763.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cmtm5

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          negative regulation of myoblast differentiation
          molecular_function
          cytokine activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080860 AK013666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080860 AY241867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080860 BC049665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080860 NM_026066.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080860 AK005375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955437 AK013666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955437 AY241867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955437 BC049665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955437 NM_026066.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955437 AK005168.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18473 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955437 AK013666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955437 AY241867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955437 BC049665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955437 NM_026066.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955437 AK005168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080860 AK013666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080860 AY241867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080860 BC049665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080860 NM_026066.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080860 AK005375.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438924 AK013666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438924 AY241867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438924 BC049665.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438924 DQ115983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438924 NM_026066.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29615174 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08117159 - -
          Whole Mouse Genome G4122A A_51_P438924 AK013666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P438924 AY241867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P438924 BC049665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P438924 DQ115983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P438924 NM_026066.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126342 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11487046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11124379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11124390 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11124388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29615139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11438362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11487050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11392584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11487061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11124374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11438363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11163588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11124378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29615183 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415134 - -
          Affymetrix HT MG-430 PM Array Plate 1429798_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1430600_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415132 - -
          Mouse Genome 430 2.0 Array 1429798_PM_s_at - -
          Mouse Genome 430 2.0 Array 1429798_s_at - -
          Mouse Genome 430 2.0 Array 1430600_PM_at - -
          Mouse Genome 430 2.0 Array 1430600_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67272_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67272_at - -
          Affymetrix Mouse Clariom S Array PSR1400005946.mm.1 TC1400000763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400005946.mm.1 NM_026066.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005946.mm.1 NM_026066.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005945.mm.1 TC1400000763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400005945.mm.1 NM_026066.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005945.mm.1 NM_026066.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005945.mm.1 NM_029390.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005945.mm.1 NM_029390.1 Refseq
          Mouse Expression Set 430 1430600_at - -
          Mouse Expression Set 430 1429798_PM_s_at - -
          Mouse Expression Set 430 1429798_s_at - -
          Mouse Expression Set 430 1430600_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430600_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429798_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430600_at - -
          Affymetrix HT Mouse Genome 430B Array 1429798_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918764 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6130390 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130390 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231274 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231274 - -
          mouse-6 v1.1 (Illumina) scl46294.4.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46294.4.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231274 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231274 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46294.4.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix