Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315090_s1
          See other GM4606 GE Assays ›
          Gene Symbol:
          Gm4606
          Gene Name:
          predicted gene 4606
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 72828606 - 72863423 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315090_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4606
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK155799.1 - 2 555 130

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream fibronectin type III domain containing 3A Fndc3a 1700094E19Rik,
          D14Ertd453e,
          F730017H24Rik,
          Fndc3,
          sys
          -130.0kb Sense Chr. 14 - 72537948 - 72710686 on NCBI Build GRCm38
          Downstream glycine cleavage system protein H (aminomethyl carrier) pseudogene Gm9199 EG668489 184.2kb Anti-sense Chr. 14 - 73025603 - 73026890 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668519 NM_001098799.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39998 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P118047 AK155799.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02883080 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02883080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11602025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11814922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11378453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11654890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29714572 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0