Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315208_m1
          See other 4930480K23RIK GE Assays ›
          Gene Symbol:
          4930480K23Rik
          Gene Name:
          RIKEN cDNA 4930480K23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 69732784 - 69767113 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315208_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930480K23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015602.1 - 1 - 2 835 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream charged multivesicular body protein 7 Chmp7 4930596K11Rik,
          6330407G04Rik,
          AI450338,
          AW550775
          -264.0 Anti-sense Chr. 14 - 69716979 - 69732570 on NCBI Build GRCm38
          Downstream tumor necrosis factor receptor superfamily,
          member 10b
          Tnfrsf10b DR5,
          KILLER,
          Ly98,
          MK,
          TRAILR2,
          TRICK2A,
          TRICK2B,
          TRICKB
          29.1kb Sense Chr. 14 - 69767472 - 69784411 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003614 AK133329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003614 AK133329.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02877970 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08152519 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08152531 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02877969 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08152537 - -
          Whole Mouse Genome G4122A A_52_P1163494 AK015602.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02877970 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08152519 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453716_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447157_PM_at - -
          Mouse Genome 430 2.0 Array 1453716_PM_at - -
          Mouse Genome 430 2.0 Array 1447157_at - -
          Mouse Genome 430 2.0 Array 1453716_at - -
          Mouse Genome 430 2.0 Array 1447157_PM_at - -
          Mouse Expression Set 430 1447157_at - -
          Mouse Expression Set 430 1447157_PM_at - -
          Mouse Expression Set 430 1453716_PM_at - -
          Mouse Expression Set 430 1453716_at - -
          Affymetrix HT Mouse Genome 430B Array 1447157_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453716_at - -
          Affymetrix HT Mouse Genome 430B Array 1447157_at - -
          Affymetrix HT Mouse Genome 430B Array 1453716_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490817 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107000300 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0