Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316205_m1
          See other KIF3C GE Assays ›
          Gene Symbol:
          Kif3c
          Gene Name:
          kinesin family member 3C
          Gene Aliases:
          mKIAA4058
          Chromosome Location:
          Chr.12: 3365116 - 3406494 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316205_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kif3c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008445.2 NP_032471.2 1 - 2 2396 86
          GenBank mRNA AF013116.1 - 1 - 2 1972 86
          AK147572.1 - 1 - 2 2198 86
          AK220283.1 - 2 - 3 2121 86
          BC052732.1 - 1 - 2 1553 86
          BC053053.1 - 1 - 2 2338 86
          BC062648.1 - 1 - 2 2071 86
          BC127063.1 - 1 - 2 1608 40109965 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000007.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kif3c

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          organelle transport along microtubule
          nucleotide binding
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP hydrolysis activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200000055.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000055.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000055.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000059.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000059.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000059.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000054.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000054.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000054.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000060.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000060.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000060.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000058.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000058.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000058.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000056.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000056.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000056.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000057.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000057.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000057.mm.1 NM_008445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000053.mm.1 TC1200000007.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000053.mm.1 NM_008445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000053.mm.1 NM_008445.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 801339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5df8fb85f4-n94h7:80/100.66.130.62:80.
          git-commit: 8f66f361c5203ebe605e386d8a840ec15b9b8304
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0