Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400004523.mm.1 |
TC1400000578.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004523.mm.1 |
NM_007447.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004523.mm.1 |
NM_007447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004523.mm.1 |
NM_201239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004523.mm.1 |
NM_201239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004529.mm.1 |
TC1400000578.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004529.mm.1 |
NM_021472.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004529.mm.1 |
NM_021472.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004529.mm.1 |
NM_201239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004529.mm.1 |
NM_201239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004524.mm.1 |
TC1400000578.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004524.mm.1 |
NM_007447.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004524.mm.1 |
NM_007447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004524.mm.1 |
NM_201239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004524.mm.1 |
NM_201239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004528.mm.1 |
TC1400000578.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004528.mm.1 |
NM_021472.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004528.mm.1 |
NM_021472.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004528.mm.1 |
NM_201239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004528.mm.1 |
NM_201239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004530.mm.1 |
TC1400000578.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004530.mm.1 |
NM_021472.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004530.mm.1 |
NM_021472.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004530.mm.1 |
NM_201239.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004530.mm.1 |
NM_201239.3 |
Refseq |