Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
TC1400002165.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_001168538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_027986.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016768.mm.1 |
NM_027986.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
TC1400002165.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_001168538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_027986.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016765.mm.1 |
NM_027986.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
TC1400002165.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_001168538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_027986.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016766.mm.1 |
NM_027986.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
TC1400002165.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_001168538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_027986.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016767.mm.1 |
NM_027986.3 |
Refseq |