Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400017663.mm.1 |
TC1400002287.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017663.mm.1 |
NR_045961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017663.mm.1 |
NR_045961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017664.mm.1 |
TC1400002287.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017664.mm.1 |
NR_045961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017664.mm.1 |
NR_045961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017666.mm.1 |
TC1400002287.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017666.mm.1 |
NR_045961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017666.mm.1 |
NR_045961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017656.mm.1 |
TC1400002287.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017656.mm.1 |
NR_045961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017656.mm.1 |
NR_045961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017665.mm.1 |
TC1400002287.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017665.mm.1 |
NR_045961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017665.mm.1 |
NR_045961.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017657.mm.1 |
TC1400002287.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017657.mm.1 |
NR_045961.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017657.mm.1 |
NR_045961.1 |
Refseq |