Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01319786_m1
          See other 1700094M23RIK GE Assays ›
          Gene Symbol:
          1700094M23Rik
          Gene Name:
          RIKEN cDNA 1700094M23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 153162834 - 153183224 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01319786_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700094M23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007071.1 - 3 - 4 534 60

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930482G09 gene 4930482G09Rik -10.6kb Sense Chr. 3 - 153147943 - 153152108 on NCBI Build GRCm38
          Downstream ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 St6galnac3 Siat7c 29.0kb Sense Chr. 3 - 153202509 - 153725133 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124939 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05125297 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05125267 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05125297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22654991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02699581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02696500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22654993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02699587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02355294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03069757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02696496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03084148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02967740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03128005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22654998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22654996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02388980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02869746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22655081 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103390372 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0