Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01320172_m1
          See other 1700109G14RIK GE Assays ›
          Gene Symbol:
          1700109G14Rik
          Gene Name:
          RIKEN cDNA 1700109G14 gene
          Gene Aliases:
          1700126G13Rik
          Chromosome Location:
          Chr.14: 61292983 - 61305334 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01320172_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700109G14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033788.1 2 - 3 589 96
          GenBank mRNA AK007149.1 - 2 - 3 589 96
          AK018935.1 - 2 - 3 552 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF106243.1 - 2 - 3 516 Chr. 14 - 61293140 - 61305260 [-] on Build GRCm38 96
          GenBankmRNA BY707180.1 - 2 - 3 552 Chr. 14 - 61293087 - 61305296 [-] on Build GRCm38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream sarcoglycan,
          gamma (dystrophin-associated glycoprotein)
          Sgcg 35kDa,
          5430420E18Rik,
          AI642964,
          gamma-SG
          -18.1kb Sense Chr. 14 - 61219106 - 61274858 on NCBI Build GRCm38
          Downstream ADP-ribosylation factor-like 11 Arl11 ARLTS1,
          C730007L20Rik
          11.3kb Anti-sense Chr. 14 - 61306288 - 61311937 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002184.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98181 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115731 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P88738 NR_033788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P88738 AK007149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P88738 AK018935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P88738 NR_033788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P88738 AK007149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P88738 AK018935.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08132411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115731 - -
          Whole Mouse Genome G4122A A_52_P88738 NR_033788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P88738 AK007149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P88738 AK018935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P88738 NR_033788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P88738 AK007149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P88738 AK018935.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110253 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11765586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29649496 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446106_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400016919.mm.1 TC1400002184.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016919.mm.1 NR_033788.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016919.mm.1 NR_033788.1 Refseq
          Mouse Expression Set 430 1446106_PM_at - -
          Mouse Expression Set 430 1446106_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 569523 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105670017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-x7z22:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0