Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282054.1 | NP_001268983.1 | 13 - 14 | 1800 | 116 | |
NM_031870.3 | NP_114076.1 | 12 - 13 | 1891 | 116 | ||
GenBank mRNA | AF178957.1 | - | 12 - 13 | 1595 | 116 | |
AF298655.1 | - | 12 - 13 | 1891 | 116 | ||
AK029837.1 | - | 13 - 14 | 1801 | 116 | ||
AK161474.1 | - | 12 - 13 | 1641 | 116 | ||
AY351585.1 | - | 12 - 13 | 1845 | 116 | ||
AY351587.1 | - | 1 - 2 | 842 | 116 | ||
AY351591.1 | - | 4 - 5 | 1232 | 116 | ||
BC138163.1 | - | 12 - 13 | 1782 | 8861182 | 116 | |
BC145838.1 | - | 12 - 13 | 1782 | 40131141 | 116 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |