Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01321033_g1
          See other PYCR2 GE Assays ›
          Gene Symbol:
          Pycr2
          Gene Name:
          pyrroline-5-carboxylate reductase family, member 2
          Gene Aliases:
          1810018M05Rik, P5cr2
          Chromosome Location:
          Chr.1: 180904274 - 180908088 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01321033_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pycr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133705.2 NP_598466.1 6 - 7 989 79
          GenBank mRNA AK007535.1 - 6 - 7 970 79
          AK150756.1 - 4 - 5 2109 79
          AK152302.1 - 4 - 5 2109 79
          AK165618.1 - 6 - 7 964 79
          AK167622.1 - 6 - 7 945 79
          BC006882.1 - 6 - 7 965 2647612 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001722.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pycr2

          Gene Ontology Categories:

          Function(s) Process(es)

          proline biosynthetic process
          cellular amino acid biosynthetic process
          pyrroline-5-carboxylate reductase activity
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878434 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878431 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK167622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK187139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 BC006882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK150756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK152302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK167622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK187139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 BC006882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK150756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249051 AK152302.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848470 AK017783.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848470 AK084890.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848470 AK087771.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848470 NM_001081112.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656145 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK167622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK187139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 BC006882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK150756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK152302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK167622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK187139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 BC006882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK150756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249051 AK152302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK167622.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK187139.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 BC006882.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK150756.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK152302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK167622.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK187139.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 BC006882.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK150756.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249051 AK152302.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20878449 - -
          Whole Mouse Genome G4122A A_51_P249051 AK167622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK187139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 BC006882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK150756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK152302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK167622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK187139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 BC006882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK150756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249051 AK152302.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131717 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00700300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00884632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00884630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20878434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20878470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01065579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00511267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00961320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00433412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00100399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00884632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00700300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01073203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00100425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00791185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00823470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20878437 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10352312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352307 - -
          Affymetrix HT MG-430 PM Array Plate 1448315_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10352306 - -
          Mouse Genome 430 2.0 Array 1448315_a_at - -
          Mouse Genome 430 2.0 Array 1448315_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69051_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69051_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100014298.mm.1 TC0100001722.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100014298.mm.1 NM_133705.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100014298.mm.1 NM_133705.2 Refseq
          Mouse Expression Set 430 1448315_a_at - -
          Mouse Expression Set 430 1448315_PM_a_at - -
          Murine Genome U74v2 Set 160339_PM_at - -
          Murine Genome U74v2 Set 160339_at - -
          Mouse Genome 430A 2.0 Array 1448315_a_at - -
          Mouse Genome 430A 2.0 Array 1448315_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448315_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448315_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 468003 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1770070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661386 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661386 - -
          mouse-6 v1.1 (Illumina) scl17133.5.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17133.5.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2998548 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2998548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0