Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
TC0700001098.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
NM_001122780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
NM_001122780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
NM_029652.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
NM_029652.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
NM_182782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009682.mm.1 |
NM_182782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
TC0700001098.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
NM_001122780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
NM_001122780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
NM_029652.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
NM_029652.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
NM_182782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009685.mm.1 |
NM_182782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
TC0700001098.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
NM_001122780.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
NM_001122780.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
NM_029652.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
NM_029652.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
NM_182782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009683.mm.1 |
NM_182782.2 |
Refseq |