Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01325962_m1
          See other APOPT1 GE Assays ›
          Gene Symbol:
          Apopt1
          Gene Name:
          apoptogenic, mitochondrial 1
          Gene Aliases:
          1700081D05Rik, 2810002N01Rik, Apop-1, Apop1
          Chromosome Location:
          Chr.12: 111713269 - 111755055 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01325962_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apopt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026511.2 NP_080787.1 4 - 5 504 92
          GenBank mRNA AK002873.1 - 4 - 5 506 92
          AK005403.1 - 4 - 5 492 92
          AK010493.1 - 4 - 5 494 92
          AK012648.1 - 4 - 5 502 92
          AK029408.1 - 4 - 5 480 92
          BC071262.1 - 4 - 5 504 6509633 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apopt1

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          positive regulation of smooth muscle cell apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of release of cytochrome c from mitochondria
          intrinsic apoptotic signaling pathway
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28745655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28745654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28745656 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060445 BC071262.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060445 BY392793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060445 AK010493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060445 AK012648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060445 AK029408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060444 BC071262.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060444 BY392793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060444 AK012648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060444 AK029408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060444 NM_026511.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563785 AK042350.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795721 AK038689.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795721 AK049160.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163004 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130865 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060444 BC071262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060444 BY392793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060444 AK012648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060444 AK029408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060444 NM_026511.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060445 BC071262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060445 BY392793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060445 AK010493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060445 AK012648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060445 AK029408.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28745666 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163004 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02608110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130865 - -
          Whole Mouse Genome G4122A A_52_P451334 AK047131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P112066 BC071262.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P112066 AK002873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P112066 AK005403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P112066 AK010493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P112066 AK012648.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122144 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163004 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09884574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116075 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28745656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28745654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28745655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28745688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10121952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10108752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28745664 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398723 - -
          Affymetrix HT MG-430 PM Array Plate 1429248_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440183_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1454309_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10398721 - -
          Mouse Genome 430 2.0 Array 1440183_x_at - -
          Mouse Genome 430 2.0 Array 1440183_PM_x_at - -
          Mouse Genome 430 2.0 Array 1429248_at - -
          Mouse Genome 430 2.0 Array 1454309_at - -
          Mouse Genome 430 2.0 Array 1454309_PM_at - -
          Mouse Genome 430 2.0 Array 1429248_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68020_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68020_at - -
          Mouse Expression Set 430 1429248_PM_at - -
          Mouse Expression Set 430 1454309_at - -
          Mouse Expression Set 430 1454309_PM_at - -
          Mouse Expression Set 430 1429248_at - -
          Murine Genome U74v2 Set 167253_PM_at - -
          Murine Genome U74v2 Set 167253_at - -
          Affymetrix HT Mouse Genome 430B Array 1454309_at - -
          Affymetrix HT Mouse Genome 430B Array 1454309_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429248_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429248_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 794902 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217827 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217827 - -
          mouse-6 v1.1 (Illumina) scl068020.5_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068020.5_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217827 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217827 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068020.5_23-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7ffdcc7b59-b9k6r:80/100.66.134.75:80.
          git-commit: c6f4609997b4833aba2fd3ad806738f0433cd95c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc