Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600018136.mm.1 |
TC0600002323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018136.mm.1 |
NM_001171007.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018136.mm.1 |
NM_001171007.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018136.mm.1 |
NM_172729.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018136.mm.1 |
NM_172729.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018137.mm.1 |
TC0600002323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018137.mm.1 |
NM_001171007.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018137.mm.1 |
NM_001171007.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018137.mm.1 |
NM_172729.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018137.mm.1 |
NM_172729.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018140.mm.1 |
TC0600002323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018140.mm.1 |
NM_172729.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018140.mm.1 |
NM_172729.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018139.mm.1 |
TC0600002323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018139.mm.1 |
NM_172729.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018139.mm.1 |
NM_172729.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018138.mm.1 |
TC0600002323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600018138.mm.1 |
NM_001171007.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600018138.mm.1 |
NM_001171007.1 |
Refseq |