Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010236.2 | NP_034366.2 | 8 - 9 | 792 | 70 | |
GenBank mRNA | AK041952.1 | - | 6 - 7 | 934 | 70 | |
AK157040.1 | - | 8 - 9 | 753 | 70 | ||
BC005484.1 | - | 8 - 9 | 735 | 3485401 | 70 | |
CF749232.1 | - | 5 - 6 | 391 | 70 | ||
U32197.1 | - | 8 - 9 | 749 | 70 | ||
U33557.1 | - | 8 - 9 | 769 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |