Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331034_g1
          See other 0610033M10RIK GE Assays ›
          Gene Symbol:
          0610033M10Rik
          Gene Name:
          RIKEN cDNA 0610033M10 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331034_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 0610033M10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK002748.1 - 1 - 2 36 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY703254.1 - 1 - 2 33 Chr. 6 - 51401060 - 51402217 [-] on Build GRCm38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 6559 Gm6559 EG625167 -8.2kb Anti-sense Chr. 6 - 51379710 - 51392791 on NCBI Build GRCm38
          Downstream nuclear factor,
          erythroid derived 2,
          like 3
          Nfe2l3 Nrf3 30.5kb Anti-sense Chr. 6 - 51432670 - 51458768 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2335933 AK002748.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05115692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05850375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05008849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05573097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05875971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05598579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05478141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24444046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432291_PM_at - -
          Mouse Genome 430 2.0 Array 1432291_PM_at - -
          Mouse Genome 430 2.0 Array 1432291_at - -
          Mouse Expression Set 430 1432291_PM_at - -
          Mouse Expression Set 430 1432291_at - -
          Affymetrix HT Mouse Genome 430B Array 1432291_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432291_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 885608 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0