Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331075_m1
          See other ORC4 GE Assays ›
          Gene Symbol:
          Orc4
          Gene Name:
          origin recognition complex, subunit 4
          Gene Aliases:
          Orc4P, Orc4l, mMmORC4
          Chromosome Location:
          Chr.2: 48902754 - 48950272 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331075_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Orc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011958.3 NP_036088.3 11 - 12 1118 142
          NR_033442.1 11 - 12 1066 142
          GenBank mRNA AJ003140.1 - 11 - 12 1079 142
          AK010258.1 - 11 - 12 1102 142
          AK011550.1 - 11 - 12 1032 142
          AK076324.1 - 11 - 12 1119 142
          AK084326.1 - 11 - 12 1087 142
          AK169291.1 - 11 - 12 1102 142
          BC015072.1 - 11 - 12 1090 4212266 142
          BC019748.1 - 11 - 12 1060 5007346 142
          Y16386.1 - 10 - 11 950 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003492.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Orc4

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA replication initiation
          nucleotide binding
          DNA binding
          DNA replication origin binding
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200029803.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029803.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029803.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029804.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029804.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029804.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029796.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029796.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029800.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029800.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029800.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029805.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029805.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029805.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029801.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029801.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029801.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029799.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029799.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029799.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029806.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029806.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029806.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029807.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029807.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029807.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029798.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029798.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029798.mm.1 NR_033442.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029802.mm.1 TC0200003492.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029802.mm.1 NM_011958.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029802.mm.1 NR_033442.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0