Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
TC1200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
NM_001164314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
NM_001164314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
NM_001164488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
NM_001164488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
NM_011710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016397.mm.1 |
NM_011710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016405.mm.1 |
TC1200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016405.mm.1 |
NM_001164314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016405.mm.1 |
NM_001164314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
TC1200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
NM_001164314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
NM_001164314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
NM_001164488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
NM_001164488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
NM_011710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016401.mm.1 |
NM_011710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
TC1200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
NM_001164314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
NM_001164314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
NM_001164488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
NM_001164488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
NM_011710.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016400.mm.1 |
NM_011710.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016408.mm.1 |
TC1200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016408.mm.1 |
NM_001164314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016408.mm.1 |
NM_001164314.1 |
Refseq |