Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333518_m1
          See other 4930515L03RIK GE Assays ›
          Gene Symbol:
          4930515L03Rik
          Gene Name:
          RIKEN cDNA 4930515L03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 16918678 - 17013726 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333518_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930515L03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040632.1 1 - 2 66 84
          GenBank mRNA AK015798.1 - 1 - 2 67 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream plexin domain containing 2 Plxdc2 1200007L24Rik,
          5430431D22Rik,
          AI646728,
          AU022916,
          AV231634,
          Tem7r
          -242.1kb Anti-sense Chr. 2 - 16356210 - 16755839 on NCBI Build GRCm38
          Downstream nebulette Nebl 1200007O21Rik,
          A630080F05Rik,
          BB140644,
          D830029A09Rik,
          Lnebl
          329.3kb Sense Chr. 2 - 17343022 - 17731068 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003009.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170851 AK015798.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 AK013144.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 AK032080.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 AK035291.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 AK077957.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 BC010483.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 BC113183.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 BC113790.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916938 NM_028140.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91894 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73355 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170851 AK015798.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04440496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04440352 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04440394 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04440526 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00313183 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04440501 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04440393 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04440571 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200024841.mm.1 TC0200003009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200024841.mm.1 NR_040632.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024841.mm.1 NR_040632.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024843.mm.1 TC0200003009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200024843.mm.1 NR_040632.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024843.mm.1 NR_040632.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024840.mm.1 TC0200003009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200024840.mm.1 NR_040632.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024840.mm.1 NR_040632.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024842.mm.1 TC0200003009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200024842.mm.1 NR_040632.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024842.mm.1 NR_040632.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 346783 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0