Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333537_g1
          See other SAPCD2 GE Assays ›
          Gene Symbol:
          Sapcd2
          Gene Name:
          suppressor APC domain containing 2
          Gene Aliases:
          2010317E24Rik, 6030458L21Rik, AL033337, ang
          Chromosome Location:
          Chr.2: 25372035 - 25378213 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333537_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sapcd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081085.2 NP_001074554.1 6 - 7 1540 80
          NM_001290407.1 NP_001277336.1 5 - 6 1438 80
          GenBank mRNA AB197927.1 - 5 - 6 1438 80
          AK008577.1 - 5 - 6 1135 80
          AK154143.1 - 6 - 7 1251 80
          AK163973.1 - 6 - 7 759 80
          AK164615.1 - 6 - 7 1215 80
          AK166625.1 - 6 - 7 1215 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000413.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006594 NM_001081085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AK166625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 BC070477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AK154143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AK163973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AB197927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AK166625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 BC070477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AK154143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AK163973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P472217 AB197927.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333742 AK049650.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333742 AK136947.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333742 BC053080.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333742 BC059836.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333742 NM_177681.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4966 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121821 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006594 NM_001081085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AK166625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 BC070477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AK154143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AK163973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AB197927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AK166625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 BC070477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AK154143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AK163973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P472217 AB197927.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121821 - -
          Whole Mouse Genome G4122A A_51_P472217 AK166625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 BC070477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AK154143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AK163973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AB197927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AK166625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 BC070477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AK154143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AK163973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472217 AB197927.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01173681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21071763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01473656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01627469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01173696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01627464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02266424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01279343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01369162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01473669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01173684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02266422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21071724 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470012 - -
          Affymetrix HT MG-430 PM Array Plate 1438615_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1429405_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429404_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470001 - -
          Mouse Genome 430 2.0 Array 1438615_PM_x_at - -
          Mouse Genome 430 2.0 Array 1429405_at - -
          Mouse Genome 430 2.0 Array 1429404_at - -
          Mouse Genome 430 2.0 Array 1438615_x_at - -
          Mouse Genome 430 2.0 Array 1429404_PM_at - -
          Mouse Genome 430 2.0 Array 1429405_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72080_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72080_at - -
          Affymetrix Mouse Clariom S Array PSR0200002341.mm.1 TC0200000413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002341.mm.1 NM_001081085.2 Refseq
          Mouse Expression Set 430 1429405_PM_at - -
          Mouse Expression Set 430 1429404_PM_at - -
          Mouse Expression Set 430 1429404_at - -
          Mouse Expression Set 430 1438615_PM_x_at - -
          Mouse Expression Set 430 1438615_x_at - -
          Mouse Expression Set 430 1429405_at - -
          Murine Genome U74v2 Set 169193_f_at - -
          Murine Genome U74v2 Set 169193_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429405_at - -
          Affymetrix HT Mouse Genome 430B Array 1438615_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1429404_at - -
          Affymetrix HT Mouse Genome 430B Array 1429405_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438615_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1429404_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644800 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644800 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0