Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333570_m1
          See other A230108P19RIK GE Assays ›
          Gene Symbol:
          A230108P19Rik
          Gene Name:
          RIKEN cDNA A230108P19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 6193251 - 6321611 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333570_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A230108P19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040333.1 4 - 5 411 121
          GenBank mRNA AK039218.1 - 4 - 5 412 121
          AK044786.1 - 4 - 5 412 121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB642882.1 - 4 - 5 412 Chr. 2 - 6193251 - 6313091 [+] on Build GRCm38 121
          GenBankmRNA BB633076.1 - 4 - 5 412 Chr. 2 - 6193251 - 6313114 [+] on Build GRCm38 121

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 13383 Gm13383 OTTMUSG00000011755 -20.3kb Anti-sense Chr. 2 - 6256247 - 6263036 on NCBI Build GRCm38
          Downstream USP6 N-terminal like Usp6nl AI316785,
          RNTRE,
          TRE2NL,
          mKIAA0019
          9.6kb Sense Chr. 2 - 6322757 - 6444363 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000059.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1700002196.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716805 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376195 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 AK033082.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 AK046703.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 AK135613.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 AK143464.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 AK146676.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 AK156563.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 BC024537.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001472087.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001472530.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001472797.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001472832.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001473861.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001475453.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001475495.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001475694.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001475762.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001475779.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001476309.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001476334.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001476516.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001477605.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001478015.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001478340.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001478377.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001478477.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001478679.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001479521.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001479624.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001479755.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001480229.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001480397.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001480545.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001480743.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001480950.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_001481156.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897391 XM_618937.3 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 AK044244.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 AK044275.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 AK044316.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 BC031440.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 BC051241.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 L14935.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477101 NM_008736.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272764 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177538 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00296490 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417843 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417745 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417962 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00296497 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00296546 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417761 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04417940 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140273 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00296546 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04417761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02222377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02141682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02141688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02141695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20978275 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200000431.mm.1 TC0200000059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000431.mm.1 NR_040333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000431.mm.1 NR_040333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000432.mm.1 TC0200000059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000432.mm.1 NR_040333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000432.mm.1 NR_040333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000430.mm.1 TC0200000059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000430.mm.1 NR_040333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000430.mm.1 NR_040333.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 756440 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106770095 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0