Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335386_m1
          See other A530001N23RIK GE Assays ›
          Gene Symbol:
          A530001N23Rik
          Gene Name:
          RIKEN cDNA A530001N23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 60483921 - 60508840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335386_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A530001N23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK040640.1 - 4 - 5 878 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 5084 Gm5084 A030007L22,
          EG328264
          -267.9kb Anti-sense Chr. 13 - 60205471 - 60216018 on NCBI Build GRCm38
          Downstream death associated protein kinase 1 Dapk1 D13Ucla1,
          DAP-Kinase
          115.2kb Anti-sense Chr. 13 - 60601947 - 60763191 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101393 AK040640.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384535 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101393 AK040640.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101393 AK040640.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10657920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11068055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11068060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10777541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10487592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10643365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11019613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11074048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10976420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10657922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10524917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10531945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10657933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11068064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10939525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29085644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0