Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335796_m1
          See other 4933406D12RIK GE Assays ›
          Gene Symbol:
          4933406D12Rik
          Gene Name:
          RIKEN cDNA 4933406D12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 146542931 - 146546108 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335796_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933406D12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040502.1 3 - 4 304 90
          GenBank mRNA AK016684.1 - 3 - 4 305 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY716371.1 - 3 - 4 305 Chr. 2 - 146542931 - 146546108 [+] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Ral GTPase activating protein,
          alpha subunit 2 (catalytic)
          Ralgapa2 A230067G21Rik,
          AS250,
          BC053994,
          RGC2,
          pp250
          -32.1kb Anti-sense Chr. 2 - 146239124 - 146512222 on NCBI Build GRCm38
          Downstream predicted gene 14114 Gm14114 OTTMUSG00000015697 281.1kb Anti-sense Chr. 2 - 146827212 - 146840777 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002175.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01426723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01568448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02067951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01568442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01568453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01568447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01568445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01410201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01398511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21719233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200017228.mm.1 TC0200002175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017228.mm.1 NR_040502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017228.mm.1 NR_040502.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100110184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0