Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336348_m1
          See other CAMK1D GE Assays ›
          Gene Symbol:
          Camk1d
          Gene Name:
          calcium/calmodulin-dependent protein kinase ID
          Gene Aliases:
          A630059D12Rik, CKLiK, CaMKIdelta, E030025C11Rik, mCKLiK
          Chromosome Location:
          Chr.2: 5293457 - 5715364 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336348_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camk1d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290375.1 NP_001277304.1 2 - 3 743 96
          NM_001290376.1 NP_001277305.1 2 - 3 259 96
          NM_177343.4 NP_796317.2 2 - 3 743 96
          GenBank mRNA AK052401.1 - 2 - 3 164 96
          AK053173.1 - 2 - 3 497 96
          AK147616.1 - 2 - 3 744 96
          AK147657.1 - 2 - 3 454 96
          AK154434.1 - 2 - 3 332 96
          AK162754.1 - 2 - 3 458 96
          AK170777.1 - 2 - 3 508 96
          AY273822.1 - 2 - 3 259 96
          BC141413.1 - 2 - 3 243 9056026 96
          BC141414.1 - 2 - 3 243 9056027 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002876.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camk1d

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          inflammatory response
          nervous system development
          metabolic process
          positive regulation of neuron projection development
          phosphorylation
          positive regulation of CREB transcription factor activity
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of phagocytosis
          regulation of dendrite development
          positive regulation of respiratory burst
          regulation of granulocyte chemotaxis
          positive regulation of neutrophil chemotaxis
          nucleotide binding
          catalytic activity
          protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          calmodulin binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200023868.mm.1 TC0200002876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023868.mm.1 NM_177343.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023875.mm.1 TC0200002876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023875.mm.1 NM_177343.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023872.mm.1 TC0200002876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023872.mm.1 NM_177343.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023876.mm.1 TC0200002876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023876.mm.1 NM_177343.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023878.mm.1 TC0200002876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023878.mm.1 NM_177343.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023869.mm.1 TC0200002876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023869.mm.1 NM_177343.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 625508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0