Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336852_m1
          See other CUTAL GE Assays ›
          Gene Symbol:
          Cutal
          Gene Name:
          cutA divalent cation tolerance homolog-like
          Gene Aliases:
          D730039F16Rik
          Chromosome Location:
          Chr.2: 34874365 - 34892133 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336852_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cutal
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK021330.1 - 5 - 6 424 66
          AK033810.1 - 5 - 6 502 66
          AK085616.1 - 5 - 6 463 66

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047626 AK033810.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047626 AK085616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047626 BC021771.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047626 NM_030021.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047626 AK021330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047621 AK033810.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047621 AK085616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047621 BC021771.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047621 NM_030021.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047621 AK021330.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165484 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138994 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178140 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047626 AK033810.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047626 AK085616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047626 BC021771.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047626 NM_030021.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047626 AK021330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047621 AK033810.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047621 AK085616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047621 BC021771.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047621 NM_030021.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2047621 AK021330.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21130199 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04482864 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138994 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04482882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00342610 - -
          Whole Mouse Genome G4122A A_51_P286131 AK033810.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286131 AK085616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286131 BC021771.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286131 NM_030021.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286131 AK021330.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138994 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01382492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01382491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21130334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02244279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02029906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01885661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01885654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01841781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01238517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01885651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21130188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471607 - -
          Affymetrix HT MG-430 PM Array Plate 1430688_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1425746_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10471599 - -
          Mouse Genome 430 2.0 Array 1425746_at - -
          Mouse Genome 430 2.0 Array 1430688_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425746_PM_at - -
          Mouse Genome 430 2.0 Array 1430688_s_at - -
          Mouse Genome 430 2.0 Array 1447155_at - -
          Mouse Genome 430 2.0 Array 1447155_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77996_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77996_at - -
          Mouse Expression Set 430 1425746_at - -
          Mouse Expression Set 430 1425746_PM_at - -
          Mouse Expression Set 430 1430688_PM_s_at - -
          Mouse Expression Set 430 1430688_s_at - -
          Mouse Genome 430A 2.0 Array 1425746_PM_at - -
          Mouse Genome 430A 2.0 Array 1425746_at - -
          Affymetrix HT Mouse Genome 430A Array 1425746_at - -
          Affymetrix HT Mouse Genome 430A Array 1425746_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430688_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430688_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103870138 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233556 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231293 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231293 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2912410 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2912410 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0