Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700000909.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000909.mm.1 |
NM_021323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000919.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000919.mm.1 |
NM_021323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000921.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000921.mm.1 |
NM_021323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000922.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000922.mm.1 |
NM_021323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000916.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000916.mm.1 |
NM_021323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000923.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000923.mm.1 |
NM_021323.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000920.mm.1 |
TC0700000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000920.mm.1 |
NM_021323.3 |
Refseq |